Hydrogen generators, Precision Hydrogen series
Leverandør: PEAK SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
63-0450 63-0300 63-0200 63-0100
564-0172EA 83630 DKK
564-0172 564-0171 564-0170 564-0169
Hydrogen generators, Precision Hydrogen series
Gasgeneratorer Hydrogengeneratorer
Precision Hydrogen series hydrogen generators are suitable for carrier gas and flame gas at standard detection limits. Hydrogen purity: 99,9995%.
  • Proven PEM Technology generates hydrogen safely and reliably
  • Desiccant dryers ensure high level of purity
  • Automatic loading pump as standard
  • Maintenance limited to replacing de-ioniser cartridge and silica gel
  • Internal leak detection with automatic shutdown features
Delivery pressure: 0 to 6,9 bar