Safety shoes, lace-up, Athletic Ascender
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6243850/37 6243850/38 6243850/39 6243850/40 6243850/41 6243850/42 6243850/43 6243850/44 6243850/45 6243850/46 6243850/47
114-5498EA 1190 DKK
114-5498 114-5499 114-5500 114-5501 114-5502 114-5503 114-5504 114-5505 114-5506 114-5507 114-5508
Safety shoes, lace-up, Athletic Ascender
Sko og støvler
Safety shoes made from water resistant leather and textile upper, 3-D lining, watertight bellows plus PU/TPU sole.
  • Breathable, absorbent, fast drying Poromax® 3-D lining
  • Coolmax® insole for an optimal perspiration absorption
  • Shock-absorbent PU/TPU sole
  • Metal-free, flexible, puncture resistant FLEXIUM ™ midsole with thermal insulation
  • Light Athletic™ metal-free toecap (200 J)
  • Forefoot and heel reinforcements
  • Speed lacing system and secured lace pocket
Colour: Black/grey.
EN ISO 20345:2004; S3, CI, SRC
Certificeringer: EN ISO 20345:2004; S3, CI, SRC.