Measuring beakers with handle, PP
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1161 1160 1159 1158 1157 1156
215-4360EA 33.8 DKK
215-4360 215-4359 215-4358 215-4357 215-4356 215-4355
Measuring beakers with handle, PP
Bægre
PP, translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 min), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.