Vacuum piston pumps, WOB-L
Leverandør: Welch by Gardner Denver
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
U2581C-50 U2567C-50 U2511C-02 U2585C-50 U2534B-01
ILMVU2581C-50EA 17480 DKK
ILMVU2581C-50 ILMVU2567C-50 ILMVU2511C-02 ILMVU2585C-50 ILMVU2534B-01
Vacuum piston pumps, WOB-L
Pumper Tromlepumper
Handy utility vacuum pump is ideal for basic lab operations.
  • Economical
  • Oil free
  • Light and portable
Applications include filtration, aspiration, desiccation, replace water aspirator, vacuum drying and many more.