Tubing, silicone, platinum cured, pharma grade, Nalgene™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8600-0200 8600-0020 8600-0030 8600-0040 8600-0050 8600-0060 8600-0070 8600-0080 8600-0100 8600-0130 8600-0120 8600-0140 8600-0190 8600-0260 8600-0310 8600-3015 8600-3016 8600-3017 8600-3018 8600-3024 8600-3025 8600-3173 8600-3182 2135-5310
228-1483EA 6640 DKK
228-1483 228-1471 228-1472 228-1473 228-1474 228-1475 228-1476 228-1477 228-1478 228-1480 228-1479 228-1481 228-1482 228-1484 228-1485 228-1486 228-1487 228-1488 228-1489 228-1490 228-1491 228-1492 228-1493 NALG2135-5310
Tubing, silicone, platinum cured, pharma grade, Nalgene™
Slanger Overførselsslange
Silicone, platinum cured.
  • Can be sterilised by most methods, including autoclaving, gamma-irradiation, and EtO
  • Intended for repeated use
  • Max. Pressure at 73 °F 23 °C: 1,0342 bar
Platinum-cured silicone tubing is flexible, durable, and translucent. This high-purity tubing is ideal for a variety of transfer applications such as pharmaceutical, laboratory, and bioprocess manufacturing. Excellent elastic memory and resiliency coupled with low hysteresis and compression set ensure that fluid transfers are reliable and accurate every time.
Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 clean room.
Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 clean room.
Certificeringer: Meets USP 87 Cytotoxicity (Agar Diffusion and MEM Elution methods); USP 88 Class VI; USP 661 (Physicochemical Tests); ISO10993-3 (Bacterial Mutagenicity – AmesAssay); ISO10993-4 (Hemolysis Direct Contact), EP 3.1.9 (Silicone Elastomer for Closures and Tubing), and FDA CFR 177.2600