96-Well assay plates
Leverandør: BIOTIX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
63300105 63300102 63300103
732-2473EA 355 DKK
732-2473 BITX63300102 BITX63300103
96-Well assay plates
Mikroplader HTS mikroplader
Medical grade, virgin PP, non treated. The large well diameter of Biotix 96-well assay plates make them ideal for a broad range of applications, including compound libraries and combinatorial chemistry. Available with either V-shaped or U-shaped well bottom to suit assay needs.
  • Raised 'chimney' wells reduce cross-contamination and allow secure application of cap mats and sealing films
  • Alphanumeric grid helps with sample identification
  • Notched bottom left corner ensures correct plate orientation
Certificeringer: Conform to ANSI/SBS 1-2004 standard forMore Product Information