Tough-Spots® laser labels
Leverandør: DIVERSIFIED BIOTECH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SPOT-1100 SPOT-1000 RNBW-1000 SPOT-1500 SPOT-5000 SPOT-1200 SPOT-1300 SPOT-1400 SPOT-2000 SPOT-2100 SPOT-2200 SPOT-2300 SPOT-2400 SPOT-2500 RSPO-2000 RNBW-1100
DIVESPOT-1100EA 1790 DKK
DIVESPOT-1100 DIVESPOT-1000 DIVERNBW-1000 DIVESPOT-1500 DIVESPOT-5000 DIVESPOT-1200 DIVESPOT-1300 DIVESPOT-1400 DIVESPOT-2000 DIVESPOT-2100 DIVESPOT-2200 DIVESPOT-2300 DIVESPOT-2400 DIVESPOT-2500 DIVERSPO-2000 DIVERNBW-1100
Tough-Spots® laser labels
Etiketter
These Tough-Spots® polyolefin labels are specially top-coated to readily accept laser printing.
  • Temperature resistant from −196 to +150 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
Tough-Spots® withstand freezer temperatures down to −196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C).