Temperature indicating tubes
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
K-1253 K-1252 K-1250
391-0084EA 1130 DKK
391-0084 391-0085 KOEHK-1250
Temperature indicating tubes
Temperaturindikatorstrips
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.