Beakers, low form, PP
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
87602 87604 87606 87608 87618 87610 87612 87616 87620 87622 87624 87626 87614
213-0377EA 242 DKK
213-0377 213-0378 213-0379 213-0380 213-0385 213-0381 213-0382 213-0384 213-0386 213-0387 213-0388 213-0389 213-0383
Beakers, low form, PP
Bægre
PP, highly translucent.
  • Excellent chemical resistance
  • Cleaning up to max 60 °C is recommended to preserve marks and inscriptions
  • For autoclaving (+121 °C) beakers with an embossed scale should be selected
With printed blue scale, beaded rim and spout.
Certificeringer: ISO 7056 - 10 ml/150 ml/400 ml/600 ml/3000 ml in addition to ISO 7056