Occupational shoes, lace-up, Air Cushion, 2600/2610
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2610/39 2610/41 2600/36 2600/35 2600/37 2600/38 2600/39 2600/40 2600/41 2600/42 2600/43 2600/44 2600/45 2600/46 2600/47 2610/40 2610/42 2610/43 2610/44 2610/45 2610/46 2610/47 2610/38 2610/37 2610/36 2610/35
ABEB2610/39EA 515 DKK
ABEB2610/39 ABEB2610/41 ABEB2600/36 ABEB2600/35 ABEB2600/37 ABEB2600/38 ABEB2600/39 ABEB2600/40 ABEB2600/41 ABEB2600/42 ABEB2600/43 ABEB2600/44 ABEB2600/45 ABEB2600/46 ABEB2600/47 ABEB2610/40 ABEB2610/42 ABEB2610/43 ABEB2610/44 ABEB2610/45 ABEB2610/46 ABEB2610/47 ABEB2610/38 ABEB2610/37 ABEB2610/36 ABEB2610/35
Occupational shoes, lace-up, Air Cushion, 2600/2610
Sko og støvler
Anti-static occupational shoes made from grained leather upper, and fuel resistant replaceable insole.
  • Sanitized® treatment reduces odour and bacterial growth
  • Air cushion PU sole with high slip resistance
Colour: White or black
Colour: White or black
Certificeringer: DGUV 112-191; EN 20347:2012; O1, SRA.