ESD occupational shoes, sandals, Nature, 4080/4090
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4090/38 4080/34 4080/35 4080/36 4080/37 4080/38 4080/39 4080/40 4080/41 4080/42 4080/43 4080/44 4080/45 4080/46 4080/47 4080/48 4090/34 4090/35 4090/36 4090/37 4090/39 4090/40 4090/41 4090/42 4090/43 4090/44 4090/45 4090/46 4090/47 4090/48
114-1857EA 493 DKK
114-1857 ABEB4080/34 ABEB4080/35 ABEB4080/36 ABEB4080/37 ABEB4080/38 ABEB4080/39 ABEB4080/40 ABEB4080/41 ABEB4080/42 ABEB4080/43 ABEB4080/44 ABEB4080/45 ABEB4080/46 ABEB4080/47 ABEB4080/48 ABEB4090/34 ABEB4090/35 ABEB4090/36 ABEB4090/37 ABEB4090/39 ABEB4090/40 ABEB4090/41 ABEB4090/42 ABEB4090/43 ABEB4090/44 ABEB4090/45 ABEB4090/46 ABEB4090/47 ABEB4090/48
ESD occupational shoes, sandals, Nature, 4080/4090
Sko og støvler
ESD sandals made from smooth leather with or without adjustable heel strap and a rubber sole (resistant to fuels, oils, acids and bases).
  • Skin friendly, leather coated, cork integrated insole with toe gripper
  • Sanitized® treatment reduces odour and bacterial growth
  • Slip resistant special rubber sole with shock-absorber heel
  • Adjustable instep area with Velcro® fastening
Colour: White
Colour: White
Certificeringer: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, SRC