Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10600034 10600047 10600048 10600003 10600004 10600008 10600009 10600013 10600014 10600096 10600117 10600118 10600065 10600079 10600080
10600034.EA 3241 DKK
10600034. 10600047. 10600048. 10600003. 10600004. 10600008. 10600009. 10600013. 10600014. 10600096. 10600117. 10600118. 10600065. 10600079. 10600080.
Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose
Overførselsmembraner
Amersham™ Protran® Premium nitrocellulose blotting membranes provide excellent sensitivity, resolution and low background in fluorescence Western blotting detection applications.
  • Available in popular pre-cut sizes
  • High binding capacity
  • Protran® Premium 0,2 NC offers low auto-fluorescence across a wide range of excitation/emission wavelengths, making the membrane suitable for fluorescence-based immunodetection and multiplexing
  • Protran® Premium 0,45 NC is an unsupported, 100% pure nitrocellulose membrane suitable for proteins of a wide range of molecular weights
Amersham Protran Premium offers excellent sensitivity, resolution, and low background, especially when used with the fluorescence Amersham ECL Plex Western Blotting Detection System.
Available with a choice of pore size: Protran® Premium 0,2 NC (with pore size 0,2 µm, binding capacity 173 to 203 µg IgG/cm²), or Protran® Premium 0,45 NC (with pore size 0,45 µm, binding capacity 162 to 180 µg IgG/cm²).
For convenience, Western blotting sandwiches (10 sandwiches/package) are also available. Each sandwich consists of pre-cut Amersham™ Protran® Premium NC membranes pre-assembled with 2×3 mm Chr filter papers.