Occupational shoes, clogs, The Original, 5080/5081
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5081/36 5081/37 5081/35
ABEB5081/36EA 327 DKK
ABEB5081/36 ABEB5081/37 ABEB5081/35
Occupational shoes, clogs, The Original, 5080/5081
Sko og støvler
Anti-static clogs made from smooth leather or perforated leather upper and a sole with cross/circle profile resistant to animal fats, oil and fuel.
  • Heel strap is foldable and adjustable
  • Leather lining
  • Sanitized® treatment reduces odour and bacterial growth
Colour: White leather, green sole
Colour: White leather, green sole
Certificeringer: TÜV GS-tested; EN ISO 20347:2012; OB, SRC