pH half cells, InLab®
Leverandør: Mettler - Toledo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
51343195 51343196
662-2874EA 1820 DKK
662-2874 662-2875
pH half cells, InLab®
Elektroder pH-elektroder
pH electrode with glass body and S7 screw head. Recommended for applications where the life time of the pH electrode is significantly shorter than the reference electrode.
InLab® Mono
- Extended reference lifetime
- Glass body gives protection against corrosive and organic samples
- Detachable cable: Different cables can be connected to the S7 screw head allowing use with a wide range of meters
InLab® Mono Plus
- Extra strong glass membrane
- Maximum temperature 130 °C
- Extended reference lifetime
- Glass body gives protection against corrosive and organic samples
- Detachable cable: Different cables can be connected to the S7 screw head allowing use with a wide range of meters