Cut resistant gloves, Edge® 16-500
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16-500/7 16-500/10 16-500/8 16-500/9
ANSE16-500/7EA 738 DKK
ANSE16-500/7 ANSE16-500/10 ANSE16-500/8 ANSE16-500/9
Cut resistant gloves, Edge® 16-500
Handsker
These interlock cotton liners feature a ³/₄ dipped natural rubber latex coating and a knitwrist cuff. They provide good cut resistance.
  • Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges
  • Ergonomically designed to follow the natural shape of the hand: delivers a better fit
  • Good cushioning, good cut resistance, good comfort
Ideal for handling glass, tiles, ceramics and slippery plastics, for waste collection, oyster-farming and working in green spaces.
Certificeringer: EN 388:2016 (X141B), EN 407 (X2XXXX); Cat. III.