Universal tongs, type Hünebeck, stainless steel
Leverandør: HAMMACHER KARL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HWN313-19 HWN309-25
HAMMHWN313-19EA 873 DKK
HAMMHWN313-19 HAMMHWN309-25
Universal tongs, type Hünebeck, stainless steel
Tænger
Universal holding tongs made from corrosion resistant WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).