Inline gas filters, Click-On™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60180-801 60180-802 60180-803 60180-804 60180-805 60180-806 60180-808 60180-843
HYPE60180-801EA 1820 DKK
HYPE60180-801 HYPE60180-802 HYPE60180-803 HYPE60180-804 HYPE60180-805 HYPE60180-806 HYPE60180-808 HYPE60180-843
Inline gas filters, Click-On™
Filtre Gasfiltre
This in-line filter removes contamination from gas with easy-to-use filters. Reuseable end-fittings containing needle valves remain installed during filter changes, preventing contamination and reducing the cost of clean gas.
  • Pure gas output 99,9999% or 6,0 grade
  • No carrier gas line contamination during filter change
  • Easy and fast replacement without the need for tools
Max. pressure: 11 bar
Certificeringer: TUEV approved.