Screw top vials and closures, 8 mm
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60180-596 60180-600
HYPE60180-596EA 5850 DKK
HYPE60180-596 548-2427
Screw top vials and closures, 8 mm
Hætteglas
These vials are compatible with a broad variety of autosamplers and have a 8-425 thread finish. While maintaining the standard outer dimensions the internal volumes of these vials range from below 300 µl to 2 ml.
  • Vials are made of superior quality 33 expansion borosilicate clear (Type 1, Class A) glass
  • Pre-assembled caps and septa are convenient and minimise contamination from handling
  • Polypropylene caps are chemically inert
  • Flanged caps are ideal for many japanese autosamplers
Certificeringer: Meets all requirements of US, EU, and JPN Pharmacopia.