BioHPLC columns, DNAPac™ PA100
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
088759 088761 043010 043011 043018 088765
555-7434EA 60470 DKK
555-7434 555-7436 555-7271 555-7272 555-7273 555-7440
BioHPLC columns, DNAPac™ PA100
Kromatografikolonner
These columns feature a strong anion exchange resin for high resolution analysis and purification of synthetic and modified oligonucleotides.