Tubing kit, Upchurch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1451 1452 1453 1454 1456 1457 1320 1321 1322 1323 1324 1325 1326 1404 1412 1413 1418 1420 1421
UPCH1451EA 6380 DKK
UPCH1451 UPCH1452 UPCH1453 UPCH1454 UPCH1456 UPCH1457 UPCH1320 UPCH1321 UPCH1322 UPCH1323 UPCH1324 UPCH1325 UPCH1326 UPCH1404 UPCH1412 UPCH1413 UPCH1418 UPCH1420 UPCH1421
Tubing kit, Upchurch Scientific®
Slanger Kromatografirør
PEEK or FEP.
NanoTight™ Fittings and Sleeves are designed to connect 70 µm to 1 mm Ext. Ø capillary tubing to any standard 10 to 32 coned port normally intended for 1/16 in Ext. Ø tubing. Additionally, NanoTight Fittings can be used to connect any 1/16" O.D. tubing -- the ETFE ferrule material used in the NanoTight product line is softer than PEEK™ materials making these fittings a good candidate for connecting thin-walled semi-rigid tubing such as FEP and ETFE into 10 to 32 ports with minimal constricting the inner diameter.
NanoTight™ Tubing Sleeves connect capillary tubing into most typical 10 to 32 coned receiving ports, withhold pressures 4000 psi (275 bar), and are manufactured using FEP fluoropolymer – a highly inert, melt-processed polymer that serves as an excellent alternative to PTFE. A wide assortment of sleeves is available, ensuring the availability of a NanoTight sleeve for most applications. Many of the sleeves feature a light color tint that can help more easily identify the inner diameter for future orders. Maximum recommended continuous operating temperature of 50 °C.