Microtube racks with lid
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M-510 M-511 M-513
212-9001EA 154 DKK
212-9001 212-9002 212-9004
Microtube racks with lid
Stativer Tube Racks
PP.
  • Designed for storage of samples down to –90 °C
  • Hinged lid
  • For 1,5 and 2 ml microtubes or 2,0 ml cryotubes