Filter tips, Ovation®
Leverandør: VISTALAB TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4060-2332 4058-6133 4060-3332 4060-1333 4060-1332 4058-5332 4058-6332 4058-5133
613-4939EA 4630 DKK
613-4939 VIST4058-6133 VIST4060-3332 VIST4060-1333 VIST4060-1332 VIST4058-5332 VIST4058-6332 VIST4058-5133
Filter tips, Ovation®
Pipettespidser
These tips are made from 100% pure virgin PP. They contain a hydrophobic filter which acts as a barrier to aqueous liquids and aerosols, protecting the pipette and sample from trace amounts of carryover.