Single channel pipettes, variable volume, MLA Digital Adjustable
Leverandør: VISTALAB TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1138 1137 1136
VIST1138EA 15840 DKK
VIST1138 VIST1137 VIST1136
Single channel pipettes, variable volume, MLA Digital Adjustable
Pipetter Automatiske pipetter
Digital adjustable pipettes are an ideal choice for laboratories that perform testing requiring multiple or non standard volumes. They are made of engineered aluminum alloy and stainless steel for maximum durability and performance.
  • Three models to cover volume range of 10 to 1000 μl
  • Lower spring forces to minimise thumb muscle activity
  • Simple dial-in settings with 'click stops' provide fast and easy volume selection
  • Streamlined shape and smooth, singlestroke operation make these pipettes easy to use
All pipettes are engraved with serial numbers toMore Product Information