Pressure bulbs
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40850002 40850001
HECH40850002EA 579 DKK
HECH40850002 HECH40850001
Pressure bulbs
Buretter
simple: with 2 valves – made of soft PVC (cadmium-free)
double: with valve - made of soft PVC (cadmium-free), ball in the net made of latex