Graduated pipettes, conformity certified, partial delivery
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40545730 40545736
HECH40545730EA 1180 DKK
HECH40545730 HECH40545736
Graduated pipettes, conformity certified, partial delivery
Pipetter Pipetter med inddeling
Glass.
  • Enzyme test pipettes with long, tapered, sturdy tip
  • Precision type, for partial delivery (not calibrated to tip), calibrated to deliver (Ex),
  • Zero at top, marked main points
  • Tolerance indicated on each pipette, amber graduations