ESD safety shoes, slip-on, HACCP, Static Control, 31392
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31392/36 31392/37 31392/38 31392/39 31392/40 31392/41 31392/42 31392/43 31392/44 31392/45 31392/46 31392/47
114-5215EA 916 DKK
114-5215 114-5216 114-5217 114-5218 114-5219 114-5220 114-5221 114-5222 114-5223 114-5224 114-5225 114-5226
ESD safety shoes, slip-on, HACCP, Static Control, 31392
Sko og støvler
Metal-free ESD shoe made from microfibre, reflective material, and a nitrile sole resistant to acids, bases (cleaning agents) and fuels.
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • ASC (anti-slip contact) nitrile rubber sole
  • PU midsole
  • Composite toecap (200 J)
  • Washable at 30 °C
For use in kitchens and the food industry.
Colour: White
Certificeringer: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRA