ESD occupational shoes, clogs, Rubber, 5100/5150
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5100/37 5100/38 5100/39 5100/40 5100/41 5100/42 5100/43 5100/44 5100/45 5100/46 5100/47 5100/36 5150/36 5150/37 5150/38 5150/39 5150/40 5150/41 5150/42 5150/43 5150/44 5150/45 5150/46 5150/47
114-2181EA 464 DKK
114-2181 114-2182 114-2183 114-2184 114-2185 114-2186 114-2187 114-2188 114-2189 114-2190 114-2191 ABEB5100/36 ABEB5150/36 ABEB5150/37 ABEB5150/38 ABEB5150/39 ABEB5150/40 ABEB5150/41 ABEB5150/42 ABEB5150/43 ABEB5150/44 ABEB5150/45 ABEB5150/46 ABEB5150/47
ESD occupational shoes, clogs, Rubber, 5100/5150
Sko og støvler
ESD clogs made from smooth leather upper, a non foldable, adjustable heel strap, and a rubber sole with cross/circle profile resistant to acids, bases (cleaning agents) and fuels.
  • Leather lining
  • PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Slip resistant special rubber sole
  • Adjustable instep area
Colour: White or blue
Certificeringer: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, WRU, SRC