ESD occupational boots, Cleanroom, 3620
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3620/35 3620/36 3620/37 3620/38 3620/39 3620/40 3620/41 3620/42 3620/43 3620/44 3620/45 3620/46 3620/47
ABEB3620/35EA 599 DKK
ABEB3620/35 ABEB3620/36 ABEB3620/37 ABEB3620/38 ABEB3620/39 ABEB3620/40 ABEB3620/41 ABEB3620/42 ABEB3620/43 ABEB3620/44 ABEB3620/45 ABEB3620/46 ABEB3620/47
ESD occupational boots, Cleanroom, 3620
Sko og støvler
Cleanroom boot made from white microfibre.
  • Fixed gaitor
  • Cleanroom textile
Boots for cleanroom use.
Colour: White
Certificeringer: EN ISO 20347:2012; OB, A, E, SRA