ESD safety shoes, lace-up, Anatom, 32156/32256
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32256/52 32256/49 32256/38 32256/51 32256/37 32256/50 32256/39 32256/40 32256/41 32256/42 32256/43 32256/44 32256/45 32256/46 32256/47 32256/48 32256/36
ABEB32256/52EA 758 DKK
ABEB32256/52 ABEB32256/49 ABEB32256/38 ABEB32256/51 ABEB32256/37 ABEB32256/50 ABEB32256/39 ABEB32256/40 ABEB32256/41 ABEB32256/42 ABEB32256/43 ABEB32256/44 ABEB32256/45 ABEB32256/46 ABEB32256/47 ABEB32256/48 ABEB32256/36
ESD safety shoes, lace-up, Anatom, 32156/32256
Sko og støvler
ESD shoe made from smooth leather with reflective inserts and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Protective steel toecap (200 J)
  • Metal-free puncture resistant midsole (S3 model)
  • Scratch resistant coating in heel and toe area
Colour: Black
Colour: Black
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2 or S3, SRC