Occupational shoes, clogs, Arrow, 8400
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8400/36 8400/38 8400/41 8400/45 8400/47
ABEB8400/36EA 495 DKK
ABEB8400/36 ABEB8400/38 ABEB8400/41 ABEB8400/45 ABEB8400/47
Occupational shoes, clogs, Arrow, 8400
Sko og støvler
Anti-static clogs made from smooth leather upper with leather heel patch and non foldable, adjustable heel strap.
  • Leather lining
  • Sanitized® treatment reduces odour and bacterial growth
  • Shock-absorbent PU midsole
  • Slip resistant PU sole
  • Adjustable instep area
Ideal for use in kitchens
Colour: White.
Certificeringer: EN ISO 20347:2012; OB, A, E, FO, SRA.