ESD occupational shoes, clogs, Rubber, 5200/5250
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5200/37 5200/38 5200/39 5200/40 5200/41 5200/42 5200/43 5200/44 5200/45 5200/46 5200/47 5200/36 5250/41 5250/38 5250/42 5250/36 5250/37 5250/39 5250/40 5250/43 5250/44 5250/45 5250/46 5250/47
114-2737EA 446 DKK
114-2737 114-2738 114-2739 114-2740 114-2741 114-2742 114-2743 114-2744 114-2745 114-1953 114-1954 114-1176 114-4622 114-4619 114-4623 114-4617 114-4618 114-4620 114-4621 114-4624 114-4625 114-4626 114-4627 114-4628
ESD occupational shoes, clogs, Rubber, 5200/5250
Sko og støvler
ESD clogs made from smooth leather, a foldable, adjustable heel strap, and a rubber sole with cross/circle profile, resistant to acids, bases (cleaning agents) and fuels.
  • Sanitized® treatment reduces odour and bacterial growth
  • Non slip special rubber sole
Colour: White or blue
Certificeringer: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, A, FO, WRU, SRC.