ESD safety shoes, Velcro® fastening, X-Light, 7131030/7131035
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7131030/35 7131030/36 7131030/37 7131030/38 7131030/39 7131030/40 7131030/41 7131030/42 7131030/43 7131030/44 7131030/45 7131030/46 7131030/47 7131030/48 7131035/35 7131035/36 7131035/37 7131035/38 7131035/39 7131035/40 7131035/41 7131035/42 7131035/43 7131035/44 7131035/45 7131035/46 7131035/47 7131035/48
ABEB7131030/35EA 616 DKK
ABEB7131030/35 ABEB7131030/36 ABEB7131030/37 ABEB7131030/38 ABEB7131030/39 ABEB7131030/40 ABEB7131030/41 ABEB7131030/42 ABEB7131030/43 ABEB7131030/44 ABEB7131030/45 ABEB7131030/46 ABEB7131030/47 ABEB7131030/48 ABEB7131035/35 ABEB7131035/36 ABEB7131035/37 ABEB7131035/38 ABEB7131035/39 ABEB7131035/40 ABEB7131035/41 ABEB7131035/42 ABEB7131035/43 ABEB7131035/44 ABEB7131035/45 ABEB7131035/46 ABEB7131035/47 ABEB7131035/48
ESD safety shoes, Velcro® fastening, X-Light, 7131030/7131035
Sko og støvler
ESD safety shoe made from smooth leather, a non foldable, adjustable Velcro® heel strap and a fuel resistant PU sole.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Instep area with elastic band
Colour: White/black
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; SB, A, E, FO, SRC.