Square cover glasses, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2845-25 2845-22 2850-18 2850-22 2850-25 2855-18 2855-22 2855-25 2845-18
CORN2845-25EA 1190 DKK
CORN2845-25 CORN2845-22 CORN2850-18 CORN2850-22 CORN2850-25 CORN2855-18 CORN2855-22 CORN2855-25 CORN2845-18
Square cover glasses, Corning®
Dækglas
These square cover glasses are designed for non sterile applications.
  • Packed one ounce per pack and 10 packs per case
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.