Cut resistant gloves, ActivArmr® 80-658
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80-658/10 80-658/7 80-658/9 80-658/8 80-658/11 80-658/6
ANSE80-658/10EA 2840 DKK
ANSE80-658/10 ANSE80-658/7 ANSE80-658/9 ANSE80-658/8 ANSE80-658/11 ANSE80-658/6
Cut resistant gloves, ActivArmr® 80-658
Handsker
This glove consists of a durable liner made of stainless steel/nylon/Kevlar®/glass fiber/cotton and a ¾ dipped natural rubber coating. Thanks to its triple-layer construction (steel/glass fibre/Kevlar®) it provides extreme resistance to cuts, splinters and burrs.
  • Extra-long cuff offers extended safety for the forearm
  • Glove is ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions
  • Good heat contact resistance for handling of warm objects
  • Natural rubber crinkle finish provides a firm grip
  • Excellent washability, with no decrease in comfort or fading
Ideal for handling heavy loads, glass sheets and sharp-edged objects, recycling, sorting (glass, cans, metal pieces) and cutting of dry, painted or galvanised pieces.
Certificeringer: EN 388:2016 (XX43E), EN 407 (X2XXXX); Cat. III