SPE plates, 96-well, HyperSep™ Cyano
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60300-821 60300-822 60300-823 60300-824
HYPE60300-821EA 4110 DKK
HYPE60300-821 HYPE60300-822 HYPE60300-823 HYPE60300-824
SPE plates, 96-well, HyperSep™ Cyano
Mikroplader Solid Phase Ekstraktions plader
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano Plates are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices.
Typical applications include:
- Retention of polar compounds from hexane and oils
- Reversed phase extraction of moderately polar compounds