Solid phase extraction cartridges, HyperSep™ Cyano
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60108-745 60108-746 60108-747 60108-748 60108-750 60108-751 60108-752
HYPE60108-745EA 2970 DKK
HYPE60108-745 HYPE60108-746 HYPE60108-747 HYPE60108-748 HYPE60108-750 HYPE60108-751 HYPE60108-752
Solid phase extraction cartridges, HyperSep™ Cyano
Kromatografikolonner
Solid phase extraction cartridges with medium polarity sorbent, ideal for the isolation of polar compounds from non polar matrices.
  • Less retentive than silica or diol
  • Used for retention of polar compounds from hexane and oils
  • Suited for reversed phase extraction of moderately polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Emballering: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.