N,N-Diethylhydroxylamine ∼85% ∼8.6 mol/L in water ≥85%
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D1178-25ML
D1178-500ML
TCIAD1178-25MLEA
156
DKK
TCIAD1178-25ML
TCIAD1178-500ML
N,N-Diethylhydroxylamine ∼85% ∼8.6 mol/L in water ≥85%
N,N-Diethylhydroxylamine
Formel:
C₄H₁₁NO Molekylvægt: 89.14 g/mol Flammepunkt: 45°C |
MDL Number:
MFCD00002126 CAS-nummer: 3710-84-7 EINECS: 223-055-4 UN nummer: 1993 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Nonaqueous Titration) | 85.0 to 90.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...