2-Nitro-4-(trifluormethyl)phenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Hydroxy-3-nitrobenzotrifluoride

N0779-5G N0779-1G
TCIAN0779-5GEA 944 DKK
TCIAN0779-5G TCIAN0779-1G
2-Nitro-4-(trifluormethyl)phenol ≥98.0% (by GC, titration analysis)
2-Nitro-4-(trifluormethyl)phenol
Formel: C₇H₄F₃NO₃
Kogepunkt: 94°C
Flammepunkt: 95°C
MDL Number: MFCD00009791
CAS-nummer: 400-99-7
EINECS: 206-927-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67294fa53909637de2de6e1c0f9525a66813308&vid=c67294fa53909637de2de6e1c0f9525a66813308&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown to Dark green clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR