Tris(perfluorbutyl)amin >60.0% (by GC) for mass spectrometry

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

P0074-25G P0074-100G
TCIAP0074-25GEA 665 DKK
TCIAP0074-25G TCIAP0074-100G
Tris(perfluorbutyl)amin >60.0% (by GC) for mass spectrometry
Tris(perfluorbutyl)amin
Formel: C₁₂F₂₇N
Molekylvægt: 671.1 g/mol
Kogepunkt: 178°C
Smeltepunkt: -55
Massefylde: 1.883
MDL Number: MFCD00000436
CAS-nummer: 311-89-7
EINECS: 206-223-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb28dd04fc748a4537d6f9019d46b65d5ed3658b&vid=f62a18ece9d9baf6a95262028d4a366c27876cc7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR