(S)-(-)-2,6-Dimethyl-8-hydroxy-2-octen ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-(-)-3,7-Dimethyl-6-octen-1-ol, (S)-(-)-ß-Citronellol, (S)-(-)-Citronellol, (-)-beta-Citronellol, (-)-β-Citronellol, (S)-3,7-Dimethyloct-6-en-1-ol, beta-Rhodinol
H60174.03
H60174.06
H60174.03EA
571
DKK
H60174.03
H60174.06
(S)-(-)-2,6-Dimethyl-8-hydroxy-2-octen ≥97%
(S)-(-)-2,6-Dimethyl-8-hydroxy-2-octen
Formel:
C₁₀H₂₀O Molekylvægt: 156.27 g/mol Kogepunkt: 225…226 °C Massefylde: 0.856 Flammepunkt: 99 °C (210 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00063214 CAS-nummer: 7540-51-4 EINECS: 231-415-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...