Toluidin blå O
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride
T0571-25G
T0571-100G
TCIAT0571-25GEA
468
DKK
TCIAT0571-25G
TCIAT0571-100G
Toluidin blå O
Toluidin blå O
Formel:
C₁₅H₁₆ClN₃S Molekylvægt: 305.83 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011934 CAS-nummer: 92-31-9 EINECS: 202-146-2 Merck Index: 14,09520 |
|
Specifikationer Testresultater
Appearance | Green to Dark green powder to crystal |
Lambda max. | 623.0 to 628.0 nm (MeOH) |
Absorbance (E1%1cm) | min. 1,700 (MeOH, 623.0 to 628.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...