3-(p-Tolyl)propionsyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 3-(4-Methylphenyl)propanoic acid, 3-(4-Methylphenyl)propionic acid, 3-(p-Tolyl)propionic Acid
T2797-25G T2797-5G
TCIAT2797-25GEA 2800 DKK
TCIAT2797-25G TCIAT2797-5G
3-(p-Tolyl)propionsyre ≥98.0% (by GC, titration analysis)
3-(p-Tolyl)propionsyre
Formel: C₁₀H₁₂O₂
Molekylvægt: 164.2 g/mol
MDL Number: MFCD00016561
CAS-nummer: 1505-50-6
EINECS: 216-132-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c21e00919f74283b3e23a36198ecd9469d92050a&vid=c21e00919f74283b3e23a36198ecd9469d92050a&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C