5-(Trifluormethyl)-o-anisidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-2-anisidine, 5-(Trifluoromethyl)-2-methoxyaniline, 2-Amino-4-(trifluoromethyl)anisole, 3-Amino-4-methoxybenzotrifluoride

M2212-5G M2212-25G
TCIAM2212-5GEA 263 DKK
TCIAM2212-5G TCIAM2212-25G
5-(Trifluormethyl)-o-anisidin ≥98.0% (by GC, titration analysis)
5-(Trifluormethyl)-o-anisidin
Formel: C₈H₈F₃NO
Molekylvægt: 191.15 g/mol
Kogepunkt: 230
Smeltepunkt: 59
MDL Number: MFCD00042486
CAS-nummer: 349-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=783786d5f1227fa022e205f27619ff4c287efa39&vid=783786d5f1227fa022e205f27619ff4c287efa39&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR