4-Methoxy-3-nitropyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M2283-5G
M2283-1G
TCIAM2283-5GEA
1630
DKK
TCIAM2283-5G
TCIAM2283-1G
4-Methoxy-3-nitropyridin ≥98.0% (by GC)
4-Methoxy-3-nitropyridine
Formel:
C₆H₆N₂O₃ Kogepunkt: 127°C |
MDL Number:
MFCD00209661 CAS-nummer: 31872-62-5 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...