3-Butyn-1-ol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

B0799-25ML B0799-5ML
TCIAB0799-25MLEA 476 DKK
TCIAB0799-25ML TCIAB0799-5ML
3-Butyn-1-ol ≥97.0% (by GC)
3-Butyn-1-ol
Formel: C₄H₆O
Kogepunkt: 130°C
Smeltepunkt: –64 °C
Massefylde: 0,9262 g/cm³ (20 °C)
Flammepunkt: 36°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002955
CAS-nummer: 927-74-2
EINECS: 213-161-9
UN nummer: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9003a17af978c2645efedf63ae3e92711424374a&vid=9003a17af978c2645efedf63ae3e92711424374a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR