5-Brom-2-Fluoranilin ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 2924-09-6
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Physical Form: Solid
Melting Point: 27°
Boiling Point: 105°/12mm
Beilstein Reference: 1241528
UN No.: 2811
MDL No.: MFCD03094181
Formel:
C₆H₅BrFN Kogepunkt: 105 °C (12 mmHg) Smeltepunkt: 27 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03094181 CAS-nummer: 2924-09-6 ADR: 6.1,III |