Methyl-4-piperidinecarboxylate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M2075-25G
M2075-5G
TCIAM2075-25GEA
591
DKK
TCIAM2075-25G
TCIAM2075-5G
Methyl-4-piperidinecarboxylate ≥98.0% (by titrimetric analysis)
Methyl-4-piperidinecarboxylate
Formel:
C₇H₁₃NO₂ Kogepunkt: 93°C Flammepunkt: 89°C |
MDL Number:
MFCD00190578 CAS-nummer: 2971-79-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...