4-(Trimethylstannyl)pyridine ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T1928-1G
T1928-5G
TCIAT1928-1GEA
1300
DKK
TCIAT1928-1G
TCIAT1928-5G
4-(Trimethylstannyl)pyridine ≥97.0% (by titrimetric analysis)
4-(Trimethylstannyl)pyridine
Formel:
C₈H₁₃NSn Opbevaringstemperatur: Fryser |
MDL Number:
MFCD03844808 CAS-nummer: 59020-06-3 UN nummer: 2788 ADR: 6.1,I |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...