2-(Trimethylstannyl)pyridine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T1742-1G T1742-5G
TCIAT1742-1GEA 1120 DKK
TCIAT1742-1G TCIAT1742-5G
2-(Trimethylstannyl)pyridine ≥98.0% (by titrimetric analysis)
2-(Trimethylstannyl)pyridine
Formel: C₈H₁₃NSn
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01631312
CAS-nummer: 13737-05-8
UN nummer: 2788
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89e1d4e9f2b601109be8ab0df9fada9dab02fe55&vid=89e1d4e9f2b601109be8ab0df9fada9dab02fe55&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR