Diethoxymethylvinylsilane ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Diethoxy(methyl)(vinyl)silane
L14224.22
L14224.14
L14224.22EA
2680
DKK
L14224.22
L14224.14
Diethoxymethylvinylsilane ≥97%
Diethoxymethylvinylsilane
Purity: 97%
CAS No.: 5507-44-8
Molecular Formula: C7H16O2Si
Formula Weight: 160.29
Flash Point: 17°(62°F)
Refractive Index: 1.400
Boiling Point: 133-134°
Beilstein Reference: 970124
UN No.: 1993
MDL No.: MFCD00009067
Formel:
C₇H₁₆O₂Si Molekylvægt: 160.29 g/mol Kogepunkt: 133…134 °C Massefylde: 0.858 Flammepunkt: 17 °C (62 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009067 CAS-nummer: 5507-44-8 EINECS: 226-850-4 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...